

2,4-Difluorobiphenyl
Catalog # APOSPC2749-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:190.19 g/mol
Melting Pt:61…66 °C
Flash Pt:83 °C
Storage Temperature:Refrigerator
EINECS:253-690-2
UN:3152
ADR:9,II
:GHS07
:GHS09
CAS Number:37847-52-2
Formula:C₁₂H₈F₂
MDL Number:MFCD00042515