

α-Bromo-3,4-difluorotoluene 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:85118-01-0
:
Formula:C₇H₅BrF₂
MDL Number:MFCD00009889
:207.02 g/mol
Boiling Pt:59…61 °C/3.5mmHg
Density:1.618 g/cm³ (25 °C)
Flash Pt:79 °C
Storage Temperature:Ambient
EINECS:285-653-1
UN:3265
ADR:8,III
:GHS05
:GHS07