
α-Bromo-2,6-difluorotoluene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:85118-00-9
:
Formula:C₇H₅BrF₂
MDL Number:MFCD00000329
Melting Pt:51…55 °C
Flash Pt:113 °C
Storage Temperature:Ambient
EINECS:285-652-6
UN:3261
ADR:8,II
:207.02 g/mol
:GHS05