
2,6-Difluorobenzylamine 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:69385-30-4
:
Formula:C₇H₇F₂N
MDL Number:MFCD00010144
:143.14 g/mol
Boiling Pt:174…177 °C
Density:1.197 g/cm³ (25 °C)
Flash Pt:65 °C
Storage Temperature:Ambient
EINECS:273-983-9
UN:2735
ADR:8,III
:GHS05