
2,5-Difluorobenzylamine 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:85118-06-5
:
Formula:C₇H₇F₂N
MDL Number:MFCD00010143
:143.14 g/mol
Boiling Pt:76…78 °C/15mmHg
Density:1.222 g/cm³ (25 °C)
Flash Pt:75 °C
Storage Temperature:Ambient
EINECS:285-658-9
UN:2735
ADR:8,III
:GHS05