Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,5-Difluorobenzoyl chloride
3,5-Difluorobenzoyl chloride 98%
3,5-Difluorobenzoyl chloride 98%
:
Apollo Scientific
3,5-Difluorobenzoyl chloride 98%
:
Apollo Scientific
:
129714-97-2
:
:
C₇H₃ClF₂O
:
MFCD00010309
:
176.55 g/mol
:
173…175 °C
:
1.41 g/cm³ (25 °C)
:
69 °C
:
Ambient
:
3265
:
8,II
:
GHS05
:
GHS07