

2,5-Difluorobenzoyl chloride 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:35730-09-7
:
Formula:C₇H₃ClF₂O
MDL Number:MFCD00009929
:176.55 g/mol
Boiling Pt:92…93 °C/34mmHg
Density:1.425 g/cm³ (25 °C)
Flash Pt:59 °C
Storage Temperature:Ambient
EINECS:252-701-8
UN:2920
ADR:8+3,II
:GHS05
:GHS02