Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,5-Difluorophenylhydrazine hydrochloride
3,5-Difluorophenylhydrazine hydrochloride 97%
3,5-Difluorophenylhydrazine hydrochloride 97%
:
Apollo Scientific
3,5-Difluorophenylhydrazine hydrochloride 97%
:
Apollo Scientific
:
502496-27-7
:
134993-88-7
:
C₆H₇Cl₃N₂
:
MFCD03094171
:
213.49 g/mol
:
261…266 °C
:
GHS07
:
Ambient