Order Entry
Export
ContactUsLinkComponent
FMOC-L-3,4-difluorophenylalanine
FMOC-L-3,4-difluorophenylalanine
Catalog # APOSPC2718-1G
Supplier:  Apollo Scientific
CAS Number:  
FMOC-L-3,4-difluorophenylalanine
Catalog # APOSPC2718-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :423.42 g/mol
Storage Temperature:Ambient
CAS Number:198560-43-9
Formula:C₂₄H₁₉F₂NO₄
MDL Number:MFCD00672550