
2,5-Difluorobenzophenone 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:85068-36-6
:
Formula:C₁₃H₈F₂O
MDL Number:MFCD00009899
:218.2 g/mol
Boiling Pt:120…123 °C/0.4mmHg
Density:1.262 g/cm³ (25 °C)
Flash Pt:>110 °C
EINECS:285-298-2
:GHS07
Storage Temperature:Ambient