Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Difluorobenzophenone
2,4-Difluorobenzophenone 97%
2,4-Difluorobenzophenone 97%
:
Apollo Scientific
2,4-Difluorobenzophenone 97%
:
Apollo Scientific
:
85068-35-5
:
:
C₁₃H₈F₂O
:
MFCD00000331
:
218.2 g/mol
:
120 °C/2mmHg
:
1.261 g/cm³ (25 °C)
:
>110 °C
:
GHS07
:
Ambient