Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,2-Difluoro-3-iodobenzene
1,2-Difluoro-3-iodobenzene 97%
1,2-Difluoro-3-iodobenzene 97%
Irritant
1,2-Difluoro-3-iodobenzene 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
1,2-Difluoro-3-iodobenzene 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
64248-57-3
:
Formula:
C₆H₃F₂I
MDL Number:
MFCD08458090
:
239.9893 g/mol
:
GHS07
Storage Temperature:
Ambient