Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,5-Difluorobenzonitrile
2,5-Difluorobenzonitrile 98%
2,5-Difluorobenzonitrile 98%
:
Apollo Scientific
2,5-Difluorobenzonitrile 98%
:
Apollo Scientific
:
64248-64-2
:
:
C₇H₃F₂N
:
MFCD00001777
:
139.1 g/mol
:
188 °C
:
33…35 °C
:
77 °C
:
Ambient
:
2930
:
6.1+4.1,II
:
GHS02
:
GHS06