2,4-Difluorobenzonitrile 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:3939-09-1
:
Formula:C₇H₃F₂N
MDL Number:MFCD00009826
Boiling Pt:189 °C
Melting Pt:43…49 °C
Flash Pt:>110 °C
Storage Temperature:Ambient
EINECS:223-523-8
UN:3439
ADR:6.1,III
:139.1 g/mol
:GHS07