
2,3-Difluorobenzonitrile 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:21524-39-0
:
Formula:C₇H₃F₂N
MDL Number:MFCD00009976
Boiling Pt:188 °C
Density:1.254 g/cm³ (25 °C)
Flash Pt:75 °C
Storage Temperature:Ambient
EINECS:244-418-3
UN:3276
ADR:6.1,III
:139.1 g/mol
:GHS07