Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Difluoro-5-nitrobenzoic acid
2,4-Difluoro-5-nitrobenzoic acid 97%
2,4-Difluoro-5-nitrobenzoic acid 97%
:
Apollo Scientific
2,4-Difluoro-5-nitrobenzoic acid 97%
:
Apollo Scientific
:
153775-33-8
:
:
C₇H₃F₂NO₄
:
MFCD04115713
:
203.1 g/mol
:
125…133 °C
:
GHS07
:
Ambient