Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,4-Difluorobenzenesulphonyl chloride
3,4-Difluorobenzenesulphonyl chloride 98%
3,4-Difluorobenzenesulphonyl chloride 98%
:
Apollo Scientific
3,4-Difluorobenzenesulphonyl chloride 98%
:
Apollo Scientific
:
145758-05-0
:
:
C₆H₃ClF₂O₂S
:
MFCD00153120
:
212.6 g/mol
:
105 °C/15mmHg
:
1.586 g/cm³ (25 °C)
:
Ambient
:
3265
:
8,II
:
GHS05