
2,4-Difluorobenzenesulphonyl chloride 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:13918-92-8
:
Formula:C₆H₃ClF₂O₂S
MDL Number:MFCD00042481
:212.6 g/mol
Boiling Pt:101…102 °C/1mmHg
Density:1.581 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:3265
ADR:8,II
:GHS05