1,3-Difluorobenzene 99%
Supplier: Apollo Scientific
Jump to:
About this item
- CAS Number:372-18-9
- Formula:C₆H₄F₂
- MDL Number:MFCD00000327
- Molecular Weight:114.09 g/mol
- Boiling Pt:82 °C
- Melting Pt:–59 °C
- Density:1.163 g/cm³ (25 °C)
- Flash Pt:2 °C
- Storage Temperature:Ambient
- EINECS:206-746-5
- UN:1993
- ADR:3,II
3 Options Available Below
Product Details & Documents
- CAS Number:372-18-9
- Formula:C₆H₄F₂
- MDL Number:MFCD00000327
- Molecular Weight:114.09 g/mol
- Boiling Pt:82 °C
- Melting Pt:–59 °C
- Density:1.163 g/cm³ (25 °C)
- Flash Pt:2 °C
- Storage Temperature:Ambient
- EINECS:206-746-5
- UN:1993
- ADR:3,II



