
2,3-Difluorobenzamide 97%
Catalog # APOSPC2615M-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:157.12 g/mol
Melting Pt:114…119 °C
:GHS07
Storage Temperature:Ambient
CAS Number:18355-75-4
Formula:C₇H₅F₂NO
MDL Number:MFCD00061137