2,6-Difluorobenzaldehyde 97%
New Product
Supplier: Apollo Scientific
Jump to:
About this item
- CAS Number:437-81-0
- Formula:C₇H₄F₂O
- MDL Number:MFCD00010293
- Molecular Weight:142.1 g/mol
- Boiling Pt:82…84 °C/15mmHg
- Melting Pt:15…17 °C
- Density:1.317 g/cm³ (25 °C)
- Flash Pt:73 °C
- Storage Temperature:Ambient
3 Options Available Below
Product Details & Documents
- CAS Number:437-81-0
- Formula:C₇H₄F₂O
- MDL Number:MFCD00010293
- Molecular Weight:142.1 g/mol
- Boiling Pt:82…84 °C/15mmHg
- Melting Pt:15…17 °C
- Density:1.317 g/cm³ (25 °C)
- Flash Pt:73 °C
- Storage Temperature:Ambient



