

2,4-Difluorobenzaldehyde 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1550-35-2
:
Formula:C₇H₄F₂O
MDL Number:MFCD00010326
:142.1 g/mol
Boiling Pt:32 °C/3mmHg
Melting Pt:44257 °C
Density:1.299 g/cm³ (25 °C)
Flash Pt:55 °C
Storage Temperature:Ambient
EINECS:216-287-2
UN:1989
ADR:3,III
:GHS02
:GHS07