Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,5-Difluoroanisole
2,5-Difluoroanisole 98%
2,5-Difluoroanisole 98%
:
Apollo Scientific
2,5-Difluoroanisole 98%
:
Apollo Scientific
:
75626-17-4
:
:
C₇H₆F₂O
:
MFCD00042561
:
144.12 g/mol
:
72…73 °C/25mmHg
:
40 °C
:
Ambient
:
1993
:
3,III
:
GHS02
:
GHS07