2,4-Difluoroanisole 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:452-10-8
:
Formula:C₇H₆F₂O
MDL Number:MFCD00010731
:144.12 g/mol
Boiling Pt:53 °C/18mmHg
Density:1.24 g/cm³ (25 °C)
Flash Pt:51 °C
Storage Temperature:Ambient
UN:1993
ADR:3,III
:GHS02
:GHS07