Order Entry
Export
Orders LinkContactUsLinkComponent
 

2,4-Difluorophenol 98%

Flammable
Irritant
Supplier:   Apollo Scientific
2,4-Difluorophenol 98%
Supplier:  Apollo Scientific
Certificates
2,4-Difluorophenol 98%

5 Options Available Below

Product Details & Documents
  • CAS Number:367-27-1
  • Formula:C₆H₄F₂O
  • MDL Number:MFCD00009715
  • Molecular Weight:130.09 g/mol
  • Boiling Pt:52…53 °C/19mmHg
  • Melting Pt:22 °C
  • Density:1.362 g/cm³ (25 °C)
  • Flash Pt:57 °C
  • Storage Temperature:Ambient
  • EINECS:206-688-0
  • UN:1993
  • ADR:3,III
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 21,80 €
    21,80 €
    1 * 25 g
       21,80 €

  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 75,30 €
    75,30 €
    1 * 100 g
       75,30 €

  • Bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 369,00 €
    369,00 €
    1 * 500 g
       369,00 €

  • Bottle
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 717,00 €
    717,00 €
    1 * 1 kg
       717,00 €

  • Bottle
    5 kg
    Log in for availability
    Log in for availability
    1 * 5 kg
    1 * 5 kg - 3 560,00 €
    3 560,00 €
    1 * 5 kg
       3 560,00 €