2,5-Difluoroaniline 98%
Bottle, 100 g
Catalog # APOSPC2600-100G
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
- CAS Number:367-30-6
- Formula:C₆H₅F₂N
- MDL Number:MFCD00007651
- Molecular Weight:129.11 g/mol
- Boiling Pt:171…178 °C
- Melting Pt:41944 °C
- Density:1.288 g/cm³ (25 °C)
- Flash Pt:68 °C
- Storage Temperature:Ambient
- EINECS:206-690-1
Specifications
Specifications
- Pack type:Bottle
- Pk:100 g



