Order Entry
Export
ContactUsLinkComponent
3-Chloro-2,4-difluorobenzotrifluoride 98%
Irritant
3-Chloro-2,4-difluorobenzotrifluoride 98%
Catalog # APOSPC2597-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-Chloro-2,4-difluorobenzotrifluoride 98%
Catalog # APOSPC2597-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
Boiling Pt:131…132 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:120770-03-8
Formula:C₇H₂ClF₅