Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Difluoroaniline
2,4-Difluoroaniline 98%
2,4-Difluoroaniline 98%
:
Apollo Scientific
2,4-Difluoroaniline 98%
:
Apollo Scientific
:
367-25-9
:
:
C₆H₅F₂N
:
MFCD00007648
:
129.11 g/mol
:
170 °C/753mmHg
:
–7.5 °C
:
1.268 g/cm³ (25 °C)
:
62 °C
:
Ambient
:
206-687-5
:
2941
:
6.1,III
:
GHS06