Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,6-Dichloro-2-(trifluoromethyl)quinoline
4,6-Dichloro-2-(trifluoromethyl)quinoline 97%
4,6-Dichloro-2-(trifluoromethyl)quinoline 97%
:
Apollo Scientific
4,6-Dichloro-2-(trifluoromethyl)quinoline 97%
:
Apollo Scientific
:
18706-33-7
:
:
C₁₀H₄Cl₂F₃N
:
MFCD00276578
:
104…107 °C
:
GHS07
:
Ambient