Order Entry
Export
ContactUsLinkComponent
1,3-Dichlorotetrafluoroacetone 98%
Toxic
1,3-Dichlorotetrafluoroacetone 98%
Catalog # APOSPC2510-25G
Supplier:  Apollo Scientific
CAS Number:  
Toxic
1,3-Dichlorotetrafluoroacetone 98%
Catalog # APOSPC2510-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :198.93 g/mol
Boiling Pt:45 °C
Density:1.52 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:2810
ADR:6.1,II
 :GHS06
CAS Number:127-21-9
Formula:ClF₂CC(O)CF₂Cl
MDL Number:MFCD00000776