Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3-Dichlorooctafluorobutane
2,3-Dichlorooctafluorobutane 97%
2,3-Dichlorooctafluorobutane 97%
APOSPC2490-1G
:
Apollo Scientific
:
2,3-Dichlorooctafluorobutane 97%
APOSPC2490-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
270.94 g/mol
:
63 °C
:
–68 °C
:
1.6801 g/cm³ (25 °C)
:
GHS07
:
Ambient
:
355-20-4
:
C₄Cl₂F₈
:
MFCD00018064