
2,4-Dichloro-3-nitrobenzotrifluoride
Catalog # APOSPC2487-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:260
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient
CAS Number:203915-49-5
Formula:C₇H₂Cl₂F₃NO₂
MDL Number:MFCD00153114