
1,2-Dichlorohexafluorocyclobutane 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:356-18-3
:
Formula:C₄Cl₂F₆
MDL Number:MFCD00019259
:232.94 g/mol
Boiling Pt:59.9 °C
Melting Pt:–24.8 °C
Density:1.6447 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient