

3,5-Dichloro-4-fluorobenzotrifluoride 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:77227-81-7
:
Formula:C₇H₂Cl₂F₄
MDL Number:MFCD00068183
:232.99 g/mol
Boiling Pt:166…168 °C
Density:1.555 g/cm³ (25 °C)
Flash Pt:73 °C
Storage Temperature:Ambient
UN:2810
ADR:6.1,III
:GHS05
:GHS06