
1,4-Dichloro-2-fluorobenzene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:348-59-4
:
Formula:C₆H₃Cl₂F
MDL Number:MFCD00060656
:164.99 g/mol
Boiling Pt:168 °C
Melting Pt:4 °C
Density:1.383 g/cm³ (25 °C)
Flash Pt:65 °C
:GHS07
Storage Temperature:Ambient