Order Entry
Export
ContactUsLinkComponent
1,2-Dichloro-1,2-difluoroethylene 96%
Irritant
1,2-Dichloro-1,2-difluoroethylene 96%
Catalog # APOSPC2370-100G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1,2-Dichloro-1,2-difluoroethylene 96%
Catalog # APOSPC2370-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Cylinder
  • Pk:
    100 g
  • Seq:
    3401
  • Pk Info:
    1/4" NPT connection

Specifications

About this item

 :
 :132.92
Boiling Pt:21…22 °C/760mmHg
Density:1.494 g/cm³ (25 °C)
 :GHS07
Storage Temperature:Ambient
CAS Number:598-88-9
Formula:C₂Cl₂F₂