
1,1-Dichloro-2,2-difluoroethylene 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:79-35-6
:
Formula:C₂Cl₂F₂
MDL Number:MFCD00039338
:132.92 g/mol
Boiling Pt:19 °C
Melting Pt:–116 °C
Density:1.4385 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:3163
ADR:2.2,
:GHS07