Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,2-Dibromo-3,3,3-trifluoropropene
1,2-Dibromo-3,3,3-trifluoropropene 97%
1,2-Dibromo-3,3,3-trifluoropropene 97%
Irritant
1,2-Dibromo-3,3,3-trifluoropropene 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
1,2-Dibromo-3,3,3-trifluoropropene 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
431-22-1
:
MFCD06800547
Formula:
C₃HBr₂F₃
MDL Number:
MFCD00042236
:
253.85 g/mol
Boiling Pt:
96 °C
:
GHS07
Storage Temperature:
Ambient