Order Entry
Export
ContactUsLinkComponent
2,4-Dibromo-6-(trifluoromethyl)aniline 97%
Toxic
2,4-Dibromo-6-(trifluoromethyl)aniline 97%
Catalog # APOSPC2330CM-25G
Supplier:  Apollo Scientific
CAS Number:  
Toxic
2,4-Dibromo-6-(trifluoromethyl)aniline 97%
Catalog # APOSPC2330CM-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :318.92 g/mol
Boiling Pt:58 °C/0.7mmHg
Melting Pt:40…43 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
 :GHS06
CAS Number:71757-14-7
Formula:C₇H₄Br₂F₃N
MDL Number:MFCD00178769