
1,4-Dibromo-1,1,2,2-tetrafluorobutane 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:18599-20-7
:
Formula:BrF₂CCF₂CH₂CH₂Br
MDL Number:MFCD00039224
:287.88 g/mol
Boiling Pt:72 °C/80mmHg
Density:1.973 g/cm³ (25 °C)
:GHS07
Storage Temperature:Ambient