
4,5-Difluorobenzimidazole
Catalog # APOSPC2313-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:154.12 g/mol
Melting Pt:211…213 °C
:GHS07
Storage Temperature:Ambient
CAS Number:236736-21-3
Formula:C₇H₄F₂N₂
MDL Number:MFCD00153122