
1,4-Difluoro-2-iodobenzene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2265-92-1
:
Formula:C₆H₃F₂I
MDL Number:MFCD03094151
:239.99 g/mol
Boiling Pt:178…179 °C
Density:1.998 g/cm³ (25 °C)
Flash Pt:85 °C
:GHS07
Storage Temperature:Ambient