Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,3-Dibromotetrafluorobenzene
1,3-Dibromotetrafluorobenzene 98%
1,3-Dibromotetrafluorobenzene 98%
:
Apollo Scientific
1,3-Dibromotetrafluorobenzene 98%
:
Apollo Scientific
:
1559-87-1
:
:
C₆Br₂F₄
:
MFCD00031533
:
307.87 g/mol
:
200 °C
:
2.24 g/cm³ (25 °C)
:
92 °C
:
216-326-3
:
GHS07
:
Ambient