
2,4-Dibromo-1-fluorobenzene 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1435-53-6
:
Formula:C₆H₃Br₂F
MDL Number:MFCD00000283
:253.9 g/mol
Boiling Pt:214…216 °C
Density:2.047 g/cm³ (25 °C)
Flash Pt:92 °C
EINECS:215-860-4
:GHS07
Storage Temperature:Ambient