Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Dibromo-4-fluoroaniline
2,6-Dibromo-4-fluoroaniline 98%
2,6-Dibromo-4-fluoroaniline 98%
:
Apollo Scientific
2,6-Dibromo-4-fluoroaniline 98%
:
Apollo Scientific
:
344-18-3
:
:
C₆H₄Br₂FN
:
MFCD00041445
:
268.91 g/mol
:
63…66 °C
:
GHS07
:
Ambient