

1,2-Dibromo-4,5-difluorobenzene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:64695-78-9
:
Formula:C₆H₂Br₂F₂
MDL Number:MFCD00009890
Melting Pt:34 °C
Flash Pt:44 °C
Storage Temperature:Ambient
EINECS:265-021-1
UN:1325
ADR:4.1,III
:271.89 g/mol
:GHS02
:GHS07