Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,2-Bis(3-aminophenyl)hexafluoropropane
2,2-Bis(3-aminophenyl)hexafluoropropane 98%
2,2-Bis(3-aminophenyl)hexafluoropropane 98%
APOSPC2186-5G
:
Apollo Scientific
:
2,2-Bis(3-aminophenyl)hexafluoropropane 98%
APOSPC2186-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
334.27 g/mol
:
Ambient
:
47250-53-3
:
C₁₅H₁₂F₆N₂
:
MFCD00191619