N-(2,4-Difluorophenyl)-4,4-dimethyl-3-oxopentanamide
Catalog # APOSPC210064-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:255.27 g/mol
Storage Temperature:Ambient
CAS Number:497059-96-8
Formula:C₁₃H₁₅F₂NO₂
MDL Number:MFCD01567378