3,3-Difluorocyclobutanecarbonitrile 97%
Catalog # APOSPC201331-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:117.1 g/mol
Storage Temperature:Ambient
CAS Number:86770-80-1
Formula:C₅H₅F₂N
MDL Number:MFCD10001482