Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,4-Dibromo-5-fluoropyridine
3,4-Dibromo-5-fluoropyridine 95+%
3,4-Dibromo-5-fluoropyridine 95+%
3,4-Dibromo-5-fluoropyridine 95+%
Supplier:
Apollo Scientific
Certificates
Expand image
3,4-Dibromo-5-fluoropyridine 95+%
Supplier:
Apollo Scientific
About this item
CAS Number:
1260843-59-1
:
Formula:
C5H2Br2FN
MDL Number:
MFCD13193300
:
254.882 g/mol
Storage Temperature:
Ambient